MilliporeSigma
  • A bacterial acetyltransferase destroys plant microtubule networks and blocks secretion.
A bacterial acetyltransferase destroys plant microtubule networks and blocks secretion.
PLoS pathogens (2012-02-10)
Amy Huei-Yi Lee, Brenden Hurley, Corinna Felsensteiner, Carmen Yea, Wenzislava Ckurshumova, Verena Bartetzko, Pauline W Wang, Van Quach, Jennifer D Lewis, Yulu C Liu, Frederik Börnke, Stephane Angers, Andrew Wilde, David S Guttman, Darrell Desveaux
ABSTRACT
The eukaryotic cytoskeleton is essential for structural support and intracellular transport, and is therefore a common target of animal pathogens. However, no phytopathogenic effector has yet been demonstrated to specifically target the plant cytoskeleton. Here we show that the Pseudomonas syringae type III secreted effector HopZ1a interacts with tubulin and polymerized microtubules. We demonstrate that HopZ1a is an acetyltransferase activated by the eukaryotic co-factor phytic acid. Activated HopZ1a acetylates itself and tubulin. The conserved autoacetylation site of the YopJ / HopZ superfamily, K289, plays a critical role in both the avirulence and virulence function of HopZ1a. Furthermore, HopZ1a requires its acetyltransferase activity to cause a dramatic decrease in Arabidopsis thaliana microtubule networks, disrupt the plant secretory pathway and suppress cell wall-mediated defense. Together, this study supports the hypothesis that HopZ1a promotes virulence through cytoskeletal and secretory disruption.
Product Description
Supelco
Dextran, analytical standard, 270,000
Supelco
Dextran, analytical standard, 25,000
Supelco
Dextran, analytical standard, Set Mp 1,000-400,000
Supelco
Dextran, analytical standard, 50,000
Supelco
Dextran, analytical standard, 670,000
Supelco
Dextran, analytical standard, 150,000
Supelco
Dextran, analytical standard, 1,000
Supelco
Dextran, analytical standard, 410,000
Supelco
Dextran, analytical standard, 80,000
Sigma-Aldrich
Protease Inhibitor Cocktail, for plant cell and tissue extracts, DMSO solution
Sigma-Aldrich
Dextran, enzymatic synth.
Supelco
Dextran, analytical standard, 5,000
Supelco
Dextran, analytical standard, 12,000
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension