MilliporeSigma
  • Phosphorylation of SNAP-23 by IkappaB kinase 2 regulates mast cell degranulation.
Phosphorylation of SNAP-23 by IkappaB kinase 2 regulates mast cell degranulation.
Cell (2008-08-12)
Kotaro Suzuki, Inder M Verma
ABSTRACT
Mast cells are known to play a pivotal role in allergic diseases. Cross-linking of the high-affinity receptor for IgE (FcepsilonRI) leads to degranulation and allergic inflammation; however, the regulatory mechanisms of IgE-dependent exocytosis remain unknown. We show here that IkappaB kinase (IKK) 2 in mast cells plays critical roles in IgE-mediated anaphylaxis in vivo, and IgE-mediated degranulation in vitro, in an NF-kB-independent manner. Upon FcvarepsilonRI stimulation, IKK2 phosphorylates SNAP-23, the target membrane soluble N-ethylmaleimide-sensitive fusion factor attachment protein receptor (SNARE), and ectopic expression of a phospho-mimetic mutant of SNAP-23 partially rescued the impaired IgE-mediated degranulation in IKK2-deficient mast cells. These results suggest that IKK2 phosphorylation of SNAP-23 leads to degranulation and anaphylactic reactions. While this reaction is NF-kB-independent, we additionally show that IKK2 also regulates late-phase allergic reactions promoted by the release of proinflammatory cytokines in an NF-kB-dependent manner. The findings suggest that IKK2 is a central player in allergic reactions.
Sigma-Aldrich
Albumin from human serum, lyophilized powder, ≥95.0% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from human serum, lyophilized powder, Fatty acid free, Globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from human serum, lyophilized powder, low endotoxin
Sigma-Aldrich
Albumin from human serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from human serum, lyophilized powder, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from human serum, lyophilized powder, essentially protease free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from human serum, lyophilized powder, essentially fatty acid free
Sigma-Aldrich
Albumin from human serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)