MilliporeSigma
  • Promotion effect of the blend containing 2'-FL, OPN and DHA on oligodendrocyte progenitor cells myelination in vitro.
Promotion effect of the blend containing 2'-FL, OPN and DHA on oligodendrocyte progenitor cells myelination in vitro.
Frontiers in nutrition (2022-11-29)
Qinggang Xie, Youbo Zhang, Jinlan Zhang, Dongying Cui, Qile Zhou, Mingruo Guo
ABSTRACT
During early neurodevelopment of infant, myelination plays an essential role in brain connectivity and emergence of behavioral and cognitive function. Early life nutrition is an important factor to shape myelination and consequently cognitive appearance. To analyze the effects of additive nutrients, including 2'-fucosyllactose (2'-FL), osteopontin (OPN), docosahexaenoic acid (DHA), on neurocognitive function and brain structure, the current study evaluated the effects of different composition of breast milk nutrients on oligodendrocyte progenitor cells (OPCs) myelination with a neural primary cell model in vitro. The study showed that the three nutrients promoted the proliferation, maturation and differentiation of OPCs into mature oligodendrocytes (OLs) in each phage of the cell growth, and the effect of the nutrients blend is obviously stronger than that of the nutrient treatment alone, showing a synergistic effect in promotion of OPCs. The results of this experiment clarified the effects of 2'-FL OPN and DHA to promote myelination development of neural cells, and laid an experimental basis for further optimization of infant formula.
Sigma-Aldrich
Anti-Rabbit IgG (H+L), CF 568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (H+L), CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Myelin Associated Glycoprotein Antibody, clone 513, clone 513, Chemicon®, from mouse
Sigma-Aldrich
Anti-Neuron Cell Surface Antigen Antibody, clone A2B5-105 (7), Alexa Fluor 488, clone A2B5-105 (7), Chemicon®, from mouse
Sigma-Aldrich
bisBenzimide H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)