MilliporeSigma
  • MTHFD2 is a metabolic checkpoint controlling effector and regulatory T cell fate and function.
MTHFD2 is a metabolic checkpoint controlling effector and regulatory T cell fate and function.
Immunity (2021-11-13)
Ayaka Sugiura, Gabriela Andrejeva, Kelsey Voss, Darren R Heintzman, Xincheng Xu, Matthew Z Madden, Xiang Ye, Katherine L Beier, Nowrin U Chowdhury, Melissa M Wolf, Arissa C Young, Dalton L Greenwood, Allison E Sewell, Shailesh K Shahi, Samantha N Freedman, Alanna M Cameron, Patrik Foerch, Tim Bourne, Juan C Garcia-Canaveras, John Karijolich, Dawn C Newcomb, Ashutosh K Mangalam, Joshua D Rabinowitz, Jeffrey C Rathmell
ABSTRACT
Antigenic stimulation promotes T cell metabolic reprogramming to meet increased biosynthetic, bioenergetic, and signaling demands. We show that the one-carbon (1C) metabolism enzyme methylenetetrahydrofolate dehydrogenase 2 (MTHFD2) regulates de novo purine synthesis and signaling in activated T cells to promote proliferation and inflammatory cytokine production. In pathogenic T helper-17 (Th17) cells, MTHFD2 prevented aberrant upregulation of the transcription factor FoxP3 along with inappropriate gain of suppressive capacity. MTHFD2 deficiency also promoted regulatory T (Treg) cell differentiation. Mechanistically, MTHFD2 inhibition led to depletion of purine pools, accumulation of purine biosynthetic intermediates, and decreased nutrient sensor mTORC1 signaling. MTHFD2 was also critical to regulate DNA and histone methylation in Th17 cells. Importantly, MTHFD2 deficiency reduced disease severity in multiple in vivo inflammatory disease models. MTHFD2 is thus a metabolic checkpoint to integrate purine metabolism with pathogenic effector cell signaling and is a potential therapeutic target within 1C metabolism pathways.
Sigma-Aldrich
Hemocyanin from Megathura crenulata (keyhole limpet)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Sigma-Aldrich
Ovalbumin (323-339) (chicken, Japanese quail)
Sigma-Aldrich
LY345899, ≥95% (HPLC)
Sigma-Aldrich
Anti-phospho ULK1 Antibody (Ser777), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture