MilliporeSigma
  • Maternal and perinatal obesity induce bronchial obstruction and pulmonary hypertension via IL-6-FoxO1-axis in later life.
Maternal and perinatal obesity induce bronchial obstruction and pulmonary hypertension via IL-6-FoxO1-axis in later life.
Nature communications (2022-07-28)
Jaco Selle, Katharina Dinger, Vanessa Jentgen, Daniela Zanetti, Johannes Will, Theodoros Georgomanolis, Christina Vohlen, Rebecca Wilke, Baktybek Kojonazarov, Oleksiy Klymenko, Jasmine Mohr, Silke V Koningsbruggen-Rietschel, Christopher J Rhodes, Anna Ulrich, Dharmesh Hirani, Tim Nestler, Margarete Odenthal, Esther Mahabir, Sreenath Nayakanti, Swati Dabral, Thomas Wunderlich, James Priest, Werner Seeger, Jörg Dötsch, Soni S Pullamsetti, Miguel A Alejandre Alcazar
ABSTRACT
Obesity is a pre-disposing condition for chronic obstructive pulmonary disease, asthma, and pulmonary arterial hypertension. Accumulating evidence suggests that metabolic influences during development can determine chronic lung diseases (CLD). We demonstrate that maternal obesity causes early metabolic disorder in the offspring. Here, interleukin-6 induced bronchial and microvascular smooth muscle cell (SMC) hyperproliferation and increased airway and pulmonary vascular resistance. The key anti-proliferative transcription factor FoxO1 was inactivated via nuclear exclusion. These findings were confirmed using primary SMC treated with interleukin-6 and pharmacological FoxO1 inhibition as well as genetic FoxO1 ablation and constitutive activation. In vivo, we reproduced the structural and functional alterations in offspring of obese dams via the SMC-specific ablation of FoxO1. The reconstitution of FoxO1 using IL-6-deficient mice and pharmacological treatment did not protect against metabolic disorder but prevented SMC hyperproliferation. In human observational studies, childhood obesity was associated with reduced forced expiratory volume in 1 s/forced vital capacity ratio Z-score (used as proxy for lung function) and asthma. We conclude that the interleukin-6-FoxO1 pathway in SMC is a molecular mechanism by which perinatal obesity programs the bronchial and vascular structure and function, thereby driving CLD development. Thus, FoxO1 reconstitution provides a potential therapeutic option for preventing this metabolic programming of CLD.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
IL-1β mouse, recombinant, expressed in E. coli, untagged, >95% (SDS-PAGE)
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests
Sigma-Aldrich
Interleukin-6 from mouse, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, carrier free
Sigma-Aldrich
STAT3 Inhibitor V, Stattic, STAT3 Inhibitor V, Stattic, CAS 19983-44-9, is a cell-permeable inhibitor of STAT3 cellular function that targets the STAT3-SH2 domain and prevents its association with upstream Kinases.
Sigma-Aldrich
Foxo1 Inhibitor, AS1842856, Foxo1 Inhibitor, AS1842856, is a cell-permeable inhibitor that blocks the transcription activity of Foxo1 (IC₅₀ = 33 nM). Directly binds to the active Foxo1, but not the Ser256-phosphorylated form.
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
DAPI, for nucleic acid staining