MilliporeSigma
AMPK suppresses Th2 cell responses by repressing mTORC2.
Experimental & molecular medicine (2022-08-24)
Mahesh Pandit, Maheshwor Timilshina, Ye Gu, Suman Acharya, Yeonseok Chung, Sang-Uk Seo, Jae-Hoon Chang
ABSTRACT
Allergic inflammation is a T helper 2 (Th2) cell-driven pathophysiological phenomenon, but the mechanism by which the metabolic cascade affects Th2 cell differentiation remains unclear. In this study, we investigated the roles of AMP-activated protein kinase (AMPK) and intracellular energy sensors in Th2 cell differentiation and the pathogenesis of allergic inflammation. Accordingly, T-cell-specific AMPK or Sirtuin 1 (Sirt1)-knockout mice were subjected to allergic inflammation, and their Th2 cell responses were investigated. The results demonstrated that inducing allergic inflammation in AMPK- and Sirt1-knockout mice increased Th2 cell responses and exacerbated allergic phenotypes. Furthermore, treatment with 5-aminoimidazole-4-carboxamide ribonucleotide (AICAR), an activator of AMPK, ameliorated allergic inflammation in mice. Mechanistically, our findings revealed that AMPK repressed mechanistic target of rapamycin complex 2 (mTORC2), which downregulated the expression of suppressor of cytokine signaling 5 (SOCS5) in CD4+ T cells. In addition, the loss of AMPK signaling reduced SOCS5 expression and increased interleukin-4-STAT6-GATA3 axis-mediated Th2 cell differentiation. Finally, the T-cell-specific deletion of Rictor, a member of mTORC2, in Sirt1T-KO mice led to the reversal of allergic exacerbation to the level in control mice. Overall, our findings suggest that AMPK in CD4+ T cells inhibits the differentiation of Th2 cells by repressing mTORC2 and thus serves as a potential target for Th2 cell-associated diseases.
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
AMPK Inhibitor, Compound C, AMPK Inhibitor, Compound C, CAS 866405-64-3, is a cell-permeable compound that inhibits KDR/VEGFR2, ALK2/BMPR-I, and AMPK kinase activities (IC₅₀ = 25.1, 148, and 234.6 nM, respectively).
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder