MilliporeSigma
  • Protocadherin 15 suppresses oligodendrocyte progenitor cell proliferation and promotes motility through distinct signalling pathways.
Protocadherin 15 suppresses oligodendrocyte progenitor cell proliferation and promotes motility through distinct signalling pathways.
Communications biology (2022-06-01)
Yilan Zhen, Carlie L Cullen, Raphael Ricci, Benjamin S Summers, Sakina Rehman, Zubair M Ahmed, Antoinette Y Foster, Ben Emery, Robert Gasperini, Kaylene M Young
ABSTRACT
Oligodendrocyte progenitor cells (OPCs) express protocadherin 15 (Pcdh15), a member of the cadherin superfamily of transmembrane proteins. Little is known about the function of Pcdh15 in the central nervous system (CNS), however, Pcdh15 expression can predict glioma aggression and promote the separation of embryonic human OPCs immediately following a cell division. Herein, we show that Pcdh15 knockdown significantly increases extracellular signal-related kinase (ERK) phosphorylation and activation to enhance OPC proliferation in vitro. Furthermore, Pcdh15 knockdown elevates Cdc42-Arp2/3 signalling and impairs actin kinetics, reducing the frequency of lamellipodial extrusion and slowing filopodial withdrawal. Pcdh15 knockdown also reduces the number of processes supported by each OPC and new process generation. Our data indicate that Pcdh15 is a critical regulator of OPC proliferation and process motility, behaviours that characterise the function of these cells in the healthy CNS, and provide mechanistic insight into the role that Pcdh15 might play in glioma progression.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
ML 141, ≥98% (HPLC)
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Arp2/3 Complex Inhibitor I, CK-666, Arp2/3 Complex Inhibitor I, CK-666, CAS 442633-00-3, is a cell-permeable selective inhibitor of actin assembly mediated by actin-related protein Arp2/3 complex (IC50 = 4 uM in human).