MilliporeSigma
  • Versican promotes T helper 17 cytotoxic inflammation and impedes oligodendrocyte precursor cell remyelination.
Versican promotes T helper 17 cytotoxic inflammation and impedes oligodendrocyte precursor cell remyelination.
Nature communications (2022-05-05)
Samira Ghorbani, Emily Jelinek, Rajiv Jain, Benjamin Buehner, Cenxiao Li, Brian M Lozinski, Susobhan Sarkar, Deepak K Kaushik, Yifei Dong, Thomas N Wight, Soheila Karimi-Abdolrezaee, Geert J Schenk, Eva M Strijbis, Jeroen Geurts, Ping Zhang, Chang-Chun Ling, V Wee Yong
ABSTRACT
Remyelination failure in multiple sclerosis (MS) contributes to progression of disability. The deficient repair results from neuroinflammation and deposition of inhibitors including chondroitin sulfate proteoglycans (CSPGs). Which CSPG member is repair-inhibitory or alters local inflammation to exacerbate injury is unknown. Here, we correlate high versican-V1 expression in MS lesions with deficient premyelinating oligodendrocytes, and highlight its selective upregulation amongst CSPG members in experimental autoimmune encephalomyelitis (EAE) lesions modeling MS. In culture, purified versican-V1 inhibits oligodendrocyte precursor cells (OPCs) and promotes T helper 17 (Th17) polarization. Versican-V1-exposed Th17 cells are particularly toxic to OPCs. In NG2CreER:MAPTmGFP mice illuminating newly formed GFP+ oligodendrocytes/myelin, difluorosamine (peracetylated,4,4-difluoro-N-acetylglucosamine) treatment from peak EAE reduces lesional versican-V1 and Th17 frequency, while enhancing GFP+ profiles. We suggest that lesion-elevated versican-V1 directly impedes OPCs while it indirectly inhibits remyelination through elevating local Th17 cytotoxic neuroinflammation. We propose CSPG-lowering drugs as potential dual pronged repair and immunomodulatory therapeutics for MS.
Sigma-Aldrich
Anti-Chondroitin 4 Sulfate Antibody, clone BE-123, ascites fluid, clone BE-123, Chemicon®
Sigma-Aldrich
Thrombospondin-1 human, recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Anti-Versican Antibody, a.a. 1360-1439 of mouse versican, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Aggrecan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Versican Antibody, a.a. 535-598 of mouse versican, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Fibronectin bovine plasma, powder, BioReagent, suitable for cell culture