MilliporeSigma
  • Autism-associated protein POGZ controls ESCs and ESC neural induction by association with esBAF.
Autism-associated protein POGZ controls ESCs and ESC neural induction by association with esBAF.
Molecular autism (2022-06-02)
Xiaoyun Sun, Linxi Cheng, Yuhua Sun
ABSTRACT
The POGZ gene has been found frequently mutated in neurodevelopmental disorders (NDDs), particularly autism spectrum disorder (ASD) and intellectual disability (ID). However, little is known about its roles in embryonic stem cells (ESCs), neural development and diseases. We generated Pogz-/- ESCs and directed ESC differentiation toward a neural fate. We performed biochemistry, ChIP-seq, ATAC-seq, and bioinformatics analyses to understand the role of POGZ. We show that POGZ is required for the maintenance of ESC identity and the up-regulation of neural genes during ESC differentiation toward a neural fate. Genome-wide binding analysis shows that POGZ is primarily localized to gene promoter and enhancer regions. POGZ functions as both a transcriptional activator and repressor, and its loss leads to deregulation of differentiation genes, including neural genes. POGZ physically associates with the SWI-SNF (esBAF) chromatin remodeler complex, and together they modulate enhancer activities via epigenetic modifications such as chromatin remodeling and histone modification. During ESC neural induction, POGZ-mediated recruitment of esBAF/BRG1 and H3K27ac are important for proper expression of neural progenitor genes. The genotype and allele relevant to human neurodevelopmental disorders is heterozygous loss of function. This work is designed to study the effects of loss of POGZ function on ESCs and during ESC neural induction. Also, this work lacks of in vivo validation using animal models. The data suggest that POGZ is both a transcription factor and a genome regulator, and its loss leads to defects in neural induction and neurogenesis.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys27) Antibody, clone RM172, clone RM172, from rabbit
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.
Sigma-Aldrich
DAPI, for nucleic acid staining