MilliporeSigma
  • Guidelines for the use of flow cytometry and cell sorting in immunological studies (third edition).
Guidelines for the use of flow cytometry and cell sorting in immunological studies (third edition).
European journal of immunology (2021-12-16)
Andrea Cossarizza, Hyun-Dong Chang, Andreas Radbruch, Sergio Abrignani, Richard Addo, Mübeccel Akdis, Immanuel Andrä, Francesco Andreata, Francesco Annunziato, Eduardo Arranz, Petra Bacher, Sudipto Bari, Vincenzo Barnaba, Joana Barros-Martins, Dirk Baumjohann, Cristian G Beccaria, David Bernardo, Dominic A Boardman, Jessica Borger, Chotima Böttcher, Leonie Brockmann, Marie Burns, Dirk H Busch, Garth Cameron, Ilenia Cammarata, Antonino Cassotta, Yinshui Chang, Fernando Gabriel Chirdo, Eleni Christakou, Luka Čičin-Šain, Laura Cook, Alexandra J Corbett, Rebecca Cornelis, Lorenzo Cosmi, Martin S Davey, Sara De Biasi, Gabriele De Simone, Genny Del Zotto, Michael Delacher, Francesca Di Rosa, James Di Santo, Andreas Diefenbach, Jun Dong, Thomas Dörner, Regine J Dress, Charles-Antoine Dutertre, Sidonia B G Eckle, Pascale Eede, Maximilien Evrard, Christine S Falk, Markus Feuerer, Simon Fillatreau, Aida Fiz-Lopez, Marie Follo, Gemma A Foulds, Julia Fröbel, Nicola Gagliani, Giovanni Galletti, Anastasia Gangaev, Natalio Garbi, José Antonio Garrote, Jens Geginat, Nicholas A Gherardin, Lara Gibellini, Florent Ginhoux, Dale I Godfrey, Paola Gruarin, Claudia Haftmann, Leo Hansmann, Christopher M Harpur, Adrian C Hayday, Guido Heine, Daniela Carolina Hernández, Martin Herrmann, Oliver Hoelsken, Qing Huang, Samuel Huber, Johanna E Huber, Jochen Huehn, Michael Hundemer, William Y K Hwang, Matteo Iannacone, Sabine M Ivison, Hans-Martin Jäck, Peter K Jani, Baerbel Keller, Nina Kessler, Steven Ketelaars, Laura Knop, Jasmin Knopf, Hui-Fern Koay, Katja Kobow, Katharina Kriegsmann, H Kristyanto, Andreas Krueger, Jenny F Kuehne, Heike Kunze-Schumacher, Pia Kvistborg, Immanuel Kwok, Daniela Latorre
ABSTRACT
The third edition of Flow Cytometry Guidelines provides the key aspects to consider when performing flow cytometry experiments and includes comprehensive sections describing phenotypes and functional assays of all major human and murine immune cell subsets. Notably, the Guidelines contain helpful tables highlighting phenotypes and key differences between human and murine cells. Another useful feature of this edition is the flow cytometry analysis of clinical samples with examples of flow cytometry applications in the context of autoimmune diseases, cancers as well as acute and chronic infectious diseases. Furthermore, there are sections detailing tips, tricks and pitfalls to avoid. All sections are written and peer-reviewed by leading flow cytometry experts and immunologists, making this edition an essential and state-of-the-art handbook for basic and clinical researchers.
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Dasatinib
Roche
Liberase TM Research Grade, medium Thermolysin concentration
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Roche
DNase I, grade II, from bovine pancreas
Roche
DNase I, from bovine pancreas
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Human Serum, from human male AB plasma, USA origin, sterile-filtered
Sigma-Aldrich
DAPI, for nucleic acid staining