MilliporeSigma
  • A group B Streptococcus alpha-like protein subunit vaccine induces functionally active antibodies in humans targeting homotypic and heterotypic strains.
A group B Streptococcus alpha-like protein subunit vaccine induces functionally active antibodies in humans targeting homotypic and heterotypic strains.
Cell reports. Medicine (2022-03-05)
Andrzej Pawlowski, Jonas Lannergård, Majela Gonzalez-Miro, Duojia Cao, Sara Larsson, Jenny J Persson, Geoff Kitson, Michael Darsley, Ane Lilleøre Rom, Morten Hedegaard, Per B Fischer, Bengt Johansson-Lindbom
ABSTRACT
Maternal vaccination is a promising strategy for preventing neonatal disease caused by group B Streptococcus. The safety and immunogenicity of the prototype vaccine GBS-NN, a fusion protein consisting of the N-terminal domains of the alpha-like proteins (Alp) αC and Rib, were recently evaluated favorably in healthy adult women in a phase 1 trial. Here we demonstrate robust immunoglobulin G (IgG) and immunoglobulin A (IgA) responses against αC and Rib, as well as against the heterotypic Alp family members Alp1-Alp3. IgA and heterotypic IgG responses are more variable between subjects and correlate with pre-existing immunity. Vaccine-induced IgG mediates opsonophagocytic killing and prevents bacterial invasion of epithelial cells. Like the vaccine-induced response, naturally acquired IgG against the vaccine domains is dominated by IgG1. Consistent with the high IgG1 cross-placental transfer rate, naturally acquired IgG against both domains reaches higher concentrations in neonatal than maternal blood, as assessed in a separate group of non-vaccinated pregnant women and their babies.
Sigma-Aldrich
Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Millipore
Benzonase® Nuclease, Purity > 90%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Anti-Human IgM (μ-chain specific) F(ab′)2 fragment−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution