MilliporeSigma
  • Parallel microfluidic chemosensitivity testing on individual slice cultures.
Parallel microfluidic chemosensitivity testing on individual slice cultures.
Lab on a chip (2014-10-03)
Tim C Chang, Andrei M Mikheev, Wilson Huynh, Raymond J Monnat, Robert C Rostomily, Albert Folch
ABSTRACT
There is a critical unmet need to tailor chemotherapies to individual patients. Personalized approaches could lower treatment toxicity, improve the patient's quality of life, and ultimately reduce mortality. However, existing models of drug activity (based on tumor cells in culture or animal models) cannot accurately predict how drugs act in patients in time to inform the best possible treatment. Here we demonstrate a microfluidic device that integrates live slice cultures with an intuitive multiwell platform that allows for exposing the slices to multiple compounds at once or in sequence. We demonstrate the response of live mouse brain slices to a range of drug doses in parallel. Drug response is measured by imaging of markers for cell apoptosis and for cell death. The platform has the potential to allow for identifying the subset of therapies of greatest potential value to individual patients, on a timescale rapid enough to guide therapeutic decision-making.
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Gey′s Balanced Salt Solution, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Staurosporine solution from Streptomyces sp., Ready Made Solution, 1 mM in DMSO (100 μg/214 μL), 0.2 μm filtered
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Triton X-100
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Sigma-Aldrich
Horse Serum, Donor Herd, USA origin, Heat inactivated, sterile-filtered, suitable for cell culture