MilliporeSigma
  • Predicting the functional states of human iPSC-derived neurons with single-cell RNA-seq and electrophysiology.
Predicting the functional states of human iPSC-derived neurons with single-cell RNA-seq and electrophysiology.
Molecular psychiatry (2016-10-21)
C Bardy, M van den Hurk, B Kakaradov, J A Erwin, B N Jaeger, R V Hernandez, T Eames, A A Paucar, M Gorris, C Marchand, R Jappelli, J Barron, A K Bryant, M Kellogg, R S Lasken, B P F Rutten, H W M Steinbusch, G W Yeo, F H Gage
ABSTRACT
Human neural progenitors derived from pluripotent stem cells develop into electrophysiologically active neurons at heterogeneous rates, which can confound disease-relevant discoveries in neurology and psychiatry. By combining patch clamping, morphological and transcriptome analysis on single-human neurons in vitro, we defined a continuum of poor to highly functional electrophysiological states of differentiated neurons. The strong correlations between action potentials, synaptic activity, dendritic complexity and gene expression highlight the importance of methods for isolating functionally comparable neurons for in vitro investigations of brain disorders. Although whole-cell electrophysiology is the gold standard for functional evaluation, it often lacks the scalability required for disease modeling studies. Here, we demonstrate a multimodal machine-learning strategy to identify new molecular features that predict the physiological states of single neurons, independently of the time spent in vitro. As further proof of concept, we selected one of the potential neurophysiological biomarkers identified in this study-GDAP1L1-to isolate highly functional live human neurons in vitro.
Sigma-Aldrich
NBQX disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
SR-95531, ≥98% (HPLC), powder
Supelco
JWH-018 5-Pentanoic acid beta-D-glucuronide solution, 100 μg/mL (Acetonitrile:Water (90:10) (v/v)), certified reference material, ampule of 1 mL
Sigma-Aldrich
L-Ascorbic acid, reagent grade