MilliporeSigma
  • Deciphering the minimal quantity of mouse primary cells to undergo nephrogenesis ex vivo.
Deciphering the minimal quantity of mouse primary cells to undergo nephrogenesis ex vivo.
Developmental dynamics : an official publication of the American Association of Anatomists (2021-09-09)
Aleksandra Rak-Raszewska, Ganna Reint, Fabienne Geiger, Florence Naillat, Seppo J Vainio
ABSTRACT
Tissue organoids derived from primary cells have high potential for studying organ development and diseases in numerous organs. They recreate the morphological structure and mimic the functions of given organ while being compact in size, easy to produce, and suitable for use in various experimental setups. In this study we established the number of cells that form mouse kidney rudiments at E11.5, and generated renal organoids of various sizes from the mouse primary cells of the metanephric mesenchyme (MM). We investigated the ability of renal organoids to undergo nephrogenesis upon Wnt/ β-catenin pathway-mediated tubule induction with a GSK-3 inhibitor (BIO) or by initiation through the ureteric bud (UB). We found that 5000 cells of MM cells are necessary to successfully form renal organoids with well-structured nephrons as judged by fluorescent microscopy, transmission electron microscopy (TEM), and quantitative Polymerase Chain Reaction (qPCR). These mouse organoids also recapitulated renal secretion function in the proximal tubules. We show that a significant decrease of cells used to generate renal mouse organoids in a dissociation/re-aggregation assay, does not interfere with development, and goes toward 3Rs. This enables generation of more experimental samples with one mouse litter, limiting the number of animals used for studies.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-WT1 Antibody, clone 6F-H2, clone 6F-H2, Upstate®, from mouse
Sigma-Aldrich
BIO, ≥98% (HPLC)