MilliporeSigma
  • Activation of TGF-beta within cultured hepatocytes and in liver injury leads to intracrine signaling with expression of connective tissue growth factor.
Activation of TGF-beta within cultured hepatocytes and in liver injury leads to intracrine signaling with expression of connective tissue growth factor.
Journal of cellular and molecular medicine (2008-02-13)
Olav A Gressner, Birgit Lahme, Monika Siluschek, Katharina Rehbein, Jens Herrmann, Ralf Weiskirchen, Axel M Gressner
ABSTRACT
Recently, synthesis and secretion of connective tissue growth factor (CTGF)/CYR61/CTGF/NOV-family member 2 (CCN2) in cultures of hepatocytes were shown, which are sensitively up-regulated by exogenous TGF-beta. In this study TGF-beta-dependent CTGF/CCN2 expression in hepatocytes cultured under completely TGF-beta-free conditions was analysed by Western-blots, metabolic labelling, and CTGF-reporter gene assays. In alkaline phosphatase monoclonal anti-alkaline phosphatase complex (APAAP)-staining of cultured hepatocytes it was demonstrated that latent TGF-beta within the hepatocytes becomes rapidly detectable during culture indicating an intracellular demasking of the mature TGF-beta antigen. Subsequent signaling to theCTGF/CCN2 promoter occurs via p-Smad2, whereas p-Smad3 does not seem to be involved. Cycloheximide did not abolish the rapid immunocytochemical appearance of mature TGF-beta, but calpain inhibitors partially suppressed intracellular TGF-beta activation and subsequently CTGF up-regulation. Calpain treatment had the reverse effect. None of the inhibitors of extracellular TGF-beta signalling was effective in the reduction of spontaneous CTGF synthesis, but intracellularly acting Alk 4-/Alk 5-specific inhibitor SB-431542 was able to diminish CTGF expression. The assumption that latent intracellular TGF-beta is activated by calpains during culture-induced stress or injurious conditions in the liver in vivo was further validated by a direct effect of calpains on the activation of recombinant latent TGF-beta. In conclusion, these data are the first to suggest the possibility of intracrine TGF-beta signalling due to calpain-dependent intracellular proteolytic activation leading to transcriptional activation of CTGF/CCN2 as a TGF-beta-sensitive reporter gene. This mechanism might be deleterious for keeping long-term hepatocyte cultures due to TGF-beta-induced apoptosis and, further, might be of relevance for induction of apoptosis or epithelial-mesenchymal transition of hepatocytes in injured liver.
Sigma-Aldrich
Calpain-1, Human Erythrocytes, Calpain-1, Human Erythrocytes, is a native calpain-1. A heterodimeric cysteine proteinase with low Ca2+ requirement (EC₅₀ = 2 µM).
Sigma-Aldrich
Calpain Inhibitor III, The Calpain Inhibitor III, also referenced under CAS 88191-84-8, controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Calpain Inhibitor IV, The Calpain Inhibitor IV controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.