MilliporeSigma
  • VEGF and VEGFR2 bind to similar pH-sensitive sites on fibronectin, exposed by heparin-mediated conformational changes.
VEGF and VEGFR2 bind to similar pH-sensitive sites on fibronectin, exposed by heparin-mediated conformational changes.
The Journal of biological chemistry (2021-03-28)
Mattia Usuelli, Timmy Meyer, Raffaele Mezzenga, Maria Mitsi
ABSTRACT
Physical interactions between vascular endothelial growth factor (VEGF), a central player in blood endothelial cell biology, and fibronectin, a major fibrillar protein of the extracellular matrix, are important determinants of angiogenic activity in health and disease. Conditions signaling the need for new blood vessel growth, such as hypoxia and low extracellular pH, increase VEGF-fibronectin interactions. These interactions can be further fine-tuned through changes in the availability of the VEGF-binding sites on fibronectin, regulated by conformational changes induced by heparin and heparan sulfate chains within the extracellular matrix. These interactions may alter VEGF bioavailability, generate gradients, or alter the way VEGF is recognized by and activates its cell-surface receptors. Here, using equilibrium and kinetic studies, we discovered that fibronectin can also interact with the extracellular domain of the VEGF receptor 2 (VEGFR2). The VEGFR2-binding sites on fibronectin show great similarity to the VEGF-binding sites, as they were also exposed upon heparin-induced conformational changes in fibronectin, and the interaction was enhanced at acidic pH. Kinetic parameters and affinities for VEGF and VEGFR2 binding to fibronectin were determined by surface plasmon resonance measurements, revealing two populations of fibronectin-binding sites for each molecule. Our data also suggest that a VEGF/VEGFR2/fibronectin triple complex may be formed by VEGF or VEGFR2 first binding to fibronectin and subsequently recruiting the third binding partner. The formation of such a complex may lead to the activation of distinct angiogenic signaling pathways, offering new possibilities for clinical applications that target angiogenesis.
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg
Sigma-Aldrich
Human Plasma Fibronectin Purified Protein, from human plasma, liquid, 1 mg/mL (100 MG pack size is lyophilized), purified protein, suitable for cell culture