MilliporeSigma
  • Isolation of Human CD49f+ Astrocytes and In Vitro iPSC-Based Neurotoxicity Assays.
Isolation of Human CD49f+ Astrocytes and In Vitro iPSC-Based Neurotoxicity Assays.
STAR protocols (2020-12-31)
Lilianne Barbar, Tomasz Rusielewicz, Matthew Zimmer, Kriti Kalpana, Valentina Fossati
ABSTRACT
Given the critical roles of astrocytes in neuroinflammation and neurological diseases, models for studying human astrocyte biology are in increasing demand. Here, we present a protocol to isolate human astrocytes from induced pluripotent stem cell (iPSC)-based cultures, neural organoids, and primary tissue, using the surface marker CD49f. Moreover, we provide protocols for in vitro co-cultures of human iPSC-derived neurons and astrocytes, as well as for neurotoxicity assays that expose neurons to conditioned media from reactive astrocytes. For complete details on the use and execution of this protocol, please refer to Barbar et al. (2020).
Sigma-Aldrich
Interleukin-1α from rat, IL-1α, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
SU-5402, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Poly-L-ornithine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched