MilliporeSigma
Human lung tryptase. Purification and characterization.
The Journal of biological chemistry (1984-09-10)
T J Smith, M W Hougland, D A Johnson
ABSTRACT
Human lung tryptase, a mast cell-derived trypsin-like serine protease, has been isolated from whole human lung tissue obtained at autopsy. Increased yields from this purification process have allowed extensive characterization of the enzyme. One of the critical steps in the purification scheme is the use of a linear heparin gradient to elute active material from cellulose phosphate. Gel filtration studies in 1.0 M NaCl yielded an apparent Mr = 135,000, and subsequent electrophoresis on sodium dodecyl sulfate-polyacrylamide gels demonstrated the presence of two active species with apparent Mr = 30,900 and 31,600. Enzymatic activity was sensitive to NaCl concentrations above 0.05 M and was only 50% in 0.15 M NaCl, decreasing to 18% in 0.6 M NaCl. The effects of synthetic and natural inhibitors have also been studied, confirming the enzyme's trypsin-like characteristics and demonstrating that naturally occurring serum inhibitors are incapable of diminishing its activity. A complete amino acid analysis showed a high tryptophan content. Lastly, antisera to human lung tryptase have been generated, and the immunological identity of active fractions has been investigated as well as the localization of the enzyme to the mast cell granule by immunohistochemical staining.
Sigma-Aldrich
Tryptase from human lung, buffered aqueous solution, ≥5 units/mg protein