MilliporeSigma
  • Targeting glioblastoma signaling and metabolism with a re-purposed brain-penetrant drug.
Targeting glioblastoma signaling and metabolism with a re-purposed brain-penetrant drug.
Cell reports (2021-11-04)
Junfeng Bi, Atif Khan, Jun Tang, Aaron M Armando, Sihan Wu, Wei Zhang, Ryan C Gimple, Alex Reed, Hui Jing, Tomoyuki Koga, Ivy Tsz-Lo Wong, Yuchao Gu, Shunichiro Miki, Huijun Yang, Briana Prager, Ellis J Curtis, Derek A Wainwright, Frank B Furnari, Jeremy N Rich, Timothy F Cloughesy, Harley I Kornblum, Oswald Quehenberger, Andrey Rzhetsky, Benjamin F Cravatt, Paul S Mischel
ABSTRACT
The highly lethal brain cancer glioblastoma (GBM) poses a daunting challenge because the blood-brain barrier renders potentially druggable amplified or mutated oncoproteins relatively inaccessible. Here, we identify sphingomyelin phosphodiesterase 1 (SMPD1), an enzyme that regulates the conversion of sphingomyelin to ceramide, as an actionable drug target in GBM. We show that the highly brain-penetrant antidepressant fluoxetine potently inhibits SMPD1 activity, killing GBMs, through inhibition of epidermal growth factor receptor (EGFR) signaling and via activation of lysosomal stress. Combining fluoxetine with temozolomide, a standard of care for GBM, causes massive increases in GBM cell death and complete tumor regression in mice. Incorporation of real-world evidence from electronic medical records from insurance databases reveals significantly increased survival in GBM patients treated with fluoxetine, which was not seen in patients treated with other selective serotonin reuptake inhibitor (SSRI) antidepressants. These results nominate the repurposing of fluoxetine as a potentially safe and promising therapy for patients with GBM and suggest prospective randomized clinical trials.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep
Sigma-Aldrich
Anti-EGFR (Ab-1) Mouse mAb (528), liquid, clone 528, Calbiochem®
Sigma-Aldrich
Anti-EGFR Antibody, Upstate®, from rabbit