MilliporeSigma
  • Controlling the polarity of human gastrointestinal organoids to investigate epithelial biology and infectious diseases.
Controlling the polarity of human gastrointestinal organoids to investigate epithelial biology and infectious diseases.
Nature protocols (2021-10-20)
Julia Y Co, Mar Margalef-Català, Denise M Monack, Manuel R Amieva
ABSTRACT
Human epithelial organoids-3D spheroids derived from adult tissue stem cells-enable investigation of epithelial physiology and disease and host interactions with microorganisms, viruses and bioactive molecules. One challenge in using organoids is the difficulty in accessing the apical, or luminal, surface of the epithelium, which is enclosed within the organoid interior. This protocol describes a method we previously developed to control human and mouse organoid polarity in suspension culture such that the apical surface faces outward to the medium (apical-out organoids). Our protocol establishes apical-out polarity rapidly (24-48 h), preserves epithelial integrity, maintains secretory and absorptive functions and allows regulation of differentiation. Here, we provide a detailed description of the organoid polarity reversal method, compatible characterization assays and an example of an application of the technology-specifically the impact of host-microbe interactions on epithelial function. Control of organoid polarity expands the possibilities of organoid use in gastrointestinal and respiratory health and disease research.
Sigma-Aldrich
γ-Secretase Inhibitor IX, Gamma-Secretase Inhibitor IX - CAS 208255-80-5, is a cell-permeable inhibitor of γ-secretase (Aβtotal IC₅₀ = 115 nM, Aβ42 IC₅₀ = 200 nM).
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 4,000, (FITC:Glucose = 1:250)
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Saponin from quillaja bark, Sapogenin content ≥10 %
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
[Leu15]-Gastrin I human, ≥95% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder