MilliporeSigma
  • Optimal Maturation of the SIV-Specific CD8+ T Cell Response after Primary Infection Is Associated with Natural Control of SIV: ANRS SIC Study.
Optimal Maturation of the SIV-Specific CD8+ T Cell Response after Primary Infection Is Associated with Natural Control of SIV: ANRS SIC Study.
Cell reports (2020-09-24)
Caroline Passaes, Antoine Millet, Vincent Madelain, Valérie Monceaux, Annie David, Pierre Versmisse, Naya Sylla, Emma Gostick, Sian Llewellyn-Lacey, David A Price, Antoine Blancher, Nathalie Dereuddre-Bosquet, Delphine Desjardins, Gianfranco Pancino, Roger Le Grand, Olivier Lambotte, Michaela Müller-Trutwin, Christine Rouzioux, Jérémie Guedj, Véronique Avettand-Fenoel, Bruno Vaslin, Asier Sáez-Cirión
ABSTRACT
Highly efficient CD8+ T cells are associated with natural HIV control, but it has remained unclear how these cells are generated and maintained. We have used a macaque model of spontaneous SIVmac251 control to monitor the development of efficient CD8+ T cell responses. Our results show that SIV-specific CD8+ T cells emerge during primary infection in all animals. The ability of CD8+ T cells to suppress SIV is suboptimal in the acute phase but increases progressively in controller macaques before the establishment of sustained low-level viremia. Controller macaques develop optimal memory-like SIV-specific CD8+ T cells early after infection. In contrast, a persistently skewed differentiation phenotype characterizes memory SIV-specific CD8+ T cells in non-controller macaques. Accordingly, the phenotype of SIV-specific CD8+ T cells defined early after infection appears to favor the development of protective immunity in controllers, whereas SIV-specific CD8+ T cells in non-controllers fail to gain antiviral potency, feasibly as a consequence of early defects imprinted in the memory pool.
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat epididymal adipocytes, Type II-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
1,3-Di-o-tolylguanidine, 99%
Sigma-Aldrich
Fumitremorgin C, from Neosartorya fischeri, film
Sigma-Aldrich
N-Acetyl-D-lactosamine, ≥98% (TLC)