MilliporeSigma
  • A directional 3D neurite outgrowth model for studying motor axon biology and disease.
A directional 3D neurite outgrowth model for studying motor axon biology and disease.
Scientific reports (2021-01-24)
Xandor M Spijkers, Svetlana Pasteuning-Vuhman, Jennifa C Dorleijn, Paul Vulto, Nienke R Wevers, R Jeroen Pasterkamp
ABSTRACT
We report a method to generate a 3D motor neuron model with segregated and directed axonal outgrowth. iPSC-derived motor neurons are cultured in extracellular matrix gel in a microfluidic platform. Neurons extend their axons into an adjacent layer of gel, whereas dendrites and soma remain predominantly in the somal compartment, as verified by immunofluorescent staining. Axonal outgrowth could be precisely quantified and was shown to respond to the chemotherapeutic drug vincristine in a highly reproducible dose-dependent manner. The model was shown susceptible to excitotoxicity upon exposure with excess glutamate and showed formation of stress granules upon excess glutamate or sodium arsenite exposure, mimicking processes common in motor neuron diseases. Importantly, outgrowing axons could be attracted and repelled through a gradient of axonal guidance cues, such as semaphorins. The platform comprises 40 chips arranged underneath a microtiter plate providing both throughput and compatibility to standard laboratory equipment. The model will thus prove ideal for studying axonal biology and disease, drug discovery and regenerative medicine.
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 647 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Tau (3-repeat isoform RD3) Antibody, clone 8E6/C11, culture supernatant, clone 8E6/C11, Upstate®
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
2-Phospho-L-ascorbic acid trisodium salt, ≥95.0% (HPLC)
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Triton X-100
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Goat serum
Sigma-Aldrich
DMH1, ≥98% (HPLC)
Sigma-Aldrich
Valproic acid sodium salt, 98%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)