MilliporeSigma
  • Rapid online buffer exchange for screening of proteins, protein complexes and cell lysates by native mass spectrometry.
Rapid online buffer exchange for screening of proteins, protein complexes and cell lysates by native mass spectrometry.
Nature protocols (2020-02-02)
Zachary L VanAernum, Florian Busch, Benjamin J Jones, Mengxuan Jia, Zibo Chen, Scott E Boyken, Aniruddha Sahasrabuddhe, David Baker, Vicki H Wysocki
ABSTRACT
It is important to assess the identity and purity of proteins and protein complexes during and after protein purification to ensure that samples are of sufficient quality for further biochemical and structural characterization, as well as for use in consumer products, chemical processes and therapeutics. Native mass spectrometry (nMS) has become an important tool in protein analysis due to its ability to retain non-covalent interactions during measurements, making it possible to obtain protein structural information with high sensitivity and at high speed. Interferences from the presence of non-volatiles are typically alleviated by offline buffer exchange, which is time-consuming and difficult to automate. We provide a protocol for rapid online buffer exchange (OBE) nMS to directly screen structural features of pre-purified proteins, protein complexes or clarified cell lysates. In the liquid chromatography coupled to mass spectrometry (LC-MS) approach described in this protocol, samples in MS-incompatible conditions are injected onto a short size-exclusion chromatography column. Proteins and protein complexes are separated from small molecule non-volatile buffer components using an aqueous, non-denaturing mobile phase. Eluted proteins and protein complexes are detected by the mass spectrometer after electrospray ionization. Mass spectra can inform regarding protein sample purity and oligomerization, and additional tandem mass spectra can help to further obtain information on protein complex subunits. Information obtained by OBE nMS can be used for fast (<5 min) quality control and can further guide protein expression and purification optimization.
Sigma-Aldrich
Chaperonin 60 from Escherichia coli, >95% (SDS-PAGE), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
Hydrochloric acid, BioReagent, 36.5-38.0%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
C Reactive Protein human, recombinant, expressed in E. coli, liquid
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%