MilliporeSigma
  • Immunogenic cell death of dendritic cells following modified vaccinia virus Ankara infection enhances CD8+ T cell proliferation.
Immunogenic cell death of dendritic cells following modified vaccinia virus Ankara infection enhances CD8+ T cell proliferation.
European journal of immunology (2018-09-28)
Kim A Tappe, Ramachandramouli Budida, Metodi V Stankov, Theresa Frenz, Harshit R Shah, Asisa Volz, Gerd Sutter, Ulrich Kalinke, Georg M N Behrens
ABSTRACT
"Immunogenic cell death" (ICD) is associated with the emission of so-called damage-associated molecular patterns (DAMPs) which trigger the immune response against dead-cell associated antigens. The secretion of the DAMP, adenosine triphosphate (ATP) has been shown to be autophagy-dependent. Here, we demonstrate that Modified Vaccinia virus Ankara (MVA), a highly attenuated strain of vaccinia virus, induces both cell death and autophagy in murine bone marrow-derived dendritic cells (BMDCs), which in turn confer the (cross-)priming of OVA-specific cytotoxic T cells (OT-I cells). Additionally, we show that MVA infection leads to increased extracellular ATP (eATP) as well as intracellular ATP (iATP) levels, with the latter being influenced by the autophagy. Furthermore, we show that the increased eATP supports the proliferation of OT-I cells and inhibition of the P2RX7 receptors results in an abrogation of the proliferation. These data reveal novel mechanisms on how MVA enhances adaptive immunity in vaccine strategies.
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)