MilliporeSigma
  • SCF(Fbw7) modulates the NFkB signaling pathway by targeting NFkB2 for ubiquitination and destruction.
SCF(Fbw7) modulates the NFkB signaling pathway by targeting NFkB2 for ubiquitination and destruction.
Cell reports (2012-06-19)
Hidefumi Fukushima, Akinobu Matsumoto, Hiroyuki Inuzuka, Bo Zhai, Alan W Lau, Lixin Wan, Daming Gao, Shavali Shaik, Min Yuan, Steven P Gygi, Eijiro Jimi, John M Asara, Keiko Nakayama, Keiichi I Nakayama, Wenyi Wei
ABSTRACT
The NFkB/Rel family of proteins play critical roles in a variety of cellular processes. Thus, their physiological activation is tightly controlled. Recently, the NFkB2/p100 precursor has been characterized as the fourth IkB type of suppressor for NFkB. However, the molecular mechanism(s) underlying regulated destruction of NFkB2 remains largely unknown. Here, we report that, unlike other IkBs, ubiquitination and destruction of NFkB2 are governed by SCF(Fbw7) in a GSK3-dependent manner. In Fbw(7-/-) cells, elevated expression of NFkB2/p100 leads to a subsequent reduction in NFkB signaling pathways and elevated sensitivity to TNFa-induced cell death. Reintroducing wild-type Fbw7, but not disease-derived mutant forms of Fbw7, rescues NFkB activity. Furthermore, T cell-specific depletion of Fbw7 also leads to reduced NFkB activity and perturbed T cell differentiation. Therefore, our work identifies Fbw7 as a physiological E3 ligase controlling NFkB20s stability. It further implicates that Fbw7 might exert its tumor-suppressor function by regulating NFkB activity.
Sigma-Aldrich
Anti-Cdh1 (Ab-2) Mouse mAb (DH01), liquid, clone DH01, Calbiochem®
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid