MilliporeSigma
  • Upregulation of metastasis-associated PRL-3 initiates chordoma in zebrafish.
Upregulation of metastasis-associated PRL-3 initiates chordoma in zebrafish.
International journal of oncology (2016-02-06)
Li Li, Hongshun Shi, Mingming Zhang, Xiaoling Guo, Fang Tong, Wenliang Zhang, Junyi Zhou, Haihe Wang, Shulan Yang
ABSTRACT
The metastasis-associated phosphatase of regenerating liver-3 (PRL-3) plays multiple roles in progression of various human cancers; however, significance of its role during development has not been addressed. Here we cloned and characterized the expression pattern of zebrafish prl-3 transcript and showed that it is ubiquitiously expressed in the first 24 h of development with both maternal and zygotic expressions. The transcripts become progressively restricted to the notochord, vessels and the intestine by 96 h post-fertilization. Notably, overexpression of zebrafish Prl-3 (zPrl-3) and human PRL-3 induces notochord malformation in zebrafish. This phenotype resembles chordoma and is confirmed by associated misexpression of notochord-specific markers. Clinical significance of the PRL-3 in chordoma is strongly suggested by detection of PRL-3 antigen in clinical chordoma specimens. Collectively, our results uncovered that aberrant overexpression of PRL-3 could initiate chordoma in early development and suggest the use of PRL-3 could be used as a predictor and a therapeutic target for chordoma.
Product Description
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Roche
DIG Northern Starter Kit, suitable for Northern blotting, sufficient for 10 labeling reactions