MilliporeSigma
  • Caspase-1 induces smooth muscle cell growth in hypoxia-induced pulmonary hypertension.
Caspase-1 induces smooth muscle cell growth in hypoxia-induced pulmonary hypertension.
American journal of physiology. Lung cellular and molecular physiology (2019-03-26)
Camilla Udjus, Fadila T Cero, Bente Halvorsen, Dina Behmen, Cathrine R Carlson, Bård A Bendiksen, Emil K S Espe, Ivar Sjaastad, Else M Løberg, Arne Yndestad, Pål Aukrust, Geir Christensen, Ole H Skjønsberg, Karl-Otto Larsen
ABSTRACT
Lung diseases with hypoxia are complicated by pulmonary hypertension, leading to heart failure and death. No pharmacological treatment exists. Increased proinflammatory cytokines are found in hypoxic patients, suggesting an inflammatory pathogenesis. Caspase-1, the effector of the inflammasome, mediates inflammation through activation of the proinflammatory cytokines interleukin (IL)-18 and IL-1β. Here, we investigate inflammasome-related mechanisms that can trigger hypoxia-induced pulmonary hypertension. Our aim was to examine whether caspase-1 induces development of hypoxia-related pulmonary hypertension and is a suitable target for therapy. Wild-type (WT) and caspase-1-/- mice were exposed to 10% oxygen for 14 days. Hypoxic caspase-1-/- mice showed lower pressure and reduced muscularization in pulmonary arteries, as well as reduced right ventricular remodeling compared with WT. Smooth muscle cell (SMC) proliferation was reduced in caspase-1-deficient pulmonary arteries and in WT arteries treated with a caspase-1 inhibitor. Impaired inflammation was shown in hypoxic caspase-1-/- mice by abolished pulmonary influx of immune cells and lower levels of IL-18, IL-1β, and IL-6, which were also reduced in the medium surrounding caspase-1 abrogated pulmonary arteries. By adding IL-18 or IL-1β to caspase-1-deficient pulmonary arteries, SMC proliferation was retained. Furthermore, inhibition of both IL-6 and phosphorylated STAT3 reduced proliferation of SMC in vitro, indicating IL-18, IL-6, and STAT3 as downstream mediators of caspase-1-induced SMC proliferation in pulmonary arteries. Caspase-1 induces SMC proliferation in pulmonary arteries through the caspase-1/IL-18/IL-6/STAT3 pathway, leading to pulmonary hypertension in mice exposed to hypoxia. We propose that caspase-1 inhibition is a potential target for treatment of pulmonary hypertension.
Product Description
Roche
Cell Proliferation ELISA, BrdU (chemiluminescent)
Sigma-Aldrich
Ac-YVAD-cmk, ≥95% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-Caspase 1 Antibody, from rabbit, purified by affinity chromatography