MilliporeSigma
SRT2183 impairs ovarian cancer by facilitating autophagy.
Aging (2020-11-24)
Tingting Sun, Yanfen Hu, Weipeng He, Yuru Shang, Xiaohong Yang, Liyun Gong, Xianbin Zhang, Peng Gong, Guofen Yang
ABSTRACT
The 5-year survival rate of ovarian cancer patients is only 47%, and developing novel drugs for ovarian cancer is needed. Herein, we evaluated if and how SRT2183, a sirtuin-1 activator, impairs the ovarian cancer cells. OVCAR-3 and A2780 cells were treated with SRT2183. Cell viability was measured by cell counting kit-8 assay and clonogenic assay. Apoptosis was determined by flow cytometry with Annexin V and propidium iodide. The level of autophagy was evaluated by western blot and immunofluorescence. The activities of AKT/mTOR/70s6k and MAPK signaling pathway were measured by immunoblot. SRT2183 inhibited the growth of ovarian cancer cells, increased the accumulation of BAX, cleaved-caspase 3 and cleaved-PARP, and decreased the level of anti-apoptotic Bcl-2 and Mcl-1. SRT2183 increased the LC3II level, and enhanced the degradation of p62/SQSTM1. SRT2183 increased the formation of GFP-LC3 puncta and induced the maturation of autophagosome. Interestingly, knockdown of autophagy related 5 and 7 significantly impaired the anti-carcinoma activity of SRT2183, implying that SRT2183 impaired the ovarian cancer cells by inducing autophagy. SRT2183 decreased the accumulation of p-Akt, p-mTOR and p-70s6k, and activated the p38 MAPK signaling pathway. This indicated that Akt/mTOR/70s6k and p38 MAPK signaling pathway might be involved in the SRT2183-mediated autophagy and apoptosis.
Sigma-Aldrich
Anti-phospho-p70 S6 Kinase (Thr389) Antibody, clone 10G7.1, clone 10G7.1, from mouse
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution