MilliporeSigma
  • Microtubule poleward flux in human cells is driven by the coordinated action of four kinesins.
Microtubule poleward flux in human cells is driven by the coordinated action of four kinesins.
The EMBO journal (2020-10-20)
Yulia Steblyanko, Girish Rajendraprasad, Mariana Osswald, Susana Eibes, Ariana Jacome, Stephan Geley, António J Pereira, Helder Maiato, Marin Barisic, Yulia Steblyanko, Girish Rajendraprasad, Mariana Osswald, Susana Eibes, Ariana Jacome, Stephan Geley, António J Pereira, Helder Maiato, Marin Barisic
ABSTRACT
Mitotic spindle microtubules (MTs) undergo continuous poleward flux, whose driving force and function in humans remain unclear. Here, we combined loss-of-function screenings with analysis of MT-dynamics in human cells to investigate the molecular mechanisms underlying MT-flux. We report that kinesin-7/CENP-E at kinetochores (KTs) is the predominant driver of MT-flux in early prometaphase, while kinesin-4/KIF4A on chromosome arms facilitates MT-flux during late prometaphase and metaphase. Both these activities work in coordination with kinesin-5/EG5 and kinesin-12/KIF15, and our data suggest that the MT-flux driving force is transmitted from non-KT-MTs to KT-MTs by the MT couplers HSET and NuMA. Additionally, we found that the MT-flux rate correlates with spindle length, and this correlation depends on the establishment of stable end-on KT-MT attachments. Strikingly, we find that MT-flux is required to regulate spindle length by counteracting kinesin 13/MCAK-dependent MT-depolymerization. Thus, our study unveils the long-sought mechanism of MT-flux in human cells as relying on the coordinated action of four kinesins to compensate for MT-depolymerization and regulate spindle length.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone VIN-11-5, purified from hybridoma cell culture
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder