MilliporeSigma
  • Cytoskeleton systems contribute differently to the functional intrinsic properties of chondrospheres.
Cytoskeleton systems contribute differently to the functional intrinsic properties of chondrospheres.
Acta biomaterialia (2020-10-13)
Anna A Gryadunova, Elizaveta V Koudan, Sergey A Rodionov, F D A S Pereira, Nina Yu Meteleva, Vladimir A Kasyanov, Vladislav A Parfenov, Alexey V Kovalev, Yusef D Khesuani, Vladimir A Mironov, Elena A Bulanova
ABSTRACT
Cytoskeleton systems, actin microfilaments, microtubules (MTs) and intermediate filaments (IFs) provide the biomechanical stability and spatial organization in cells. To understand the specific contributions of each cytoskeleton systems to intrinsic properties of spheroids, we've scrutinized the effects of the cytoskeleton perturbants, cytochalasin D (Cyto D), nocodazole (Noc) and withaferin A (WFA) on fusion, spreading on adhesive surface, morphology and biomechanics of chondrospheres (CSs). We confirmed that treatment with Cyto D but not with Noc or WFA severely affected CSs fusion and spreading dynamics and significantly reduced biomechanical properties of cell aggregates. Noc treatment affected spheroids spreading but not the fusion and surprisingly enhanced their stiffness. Vimentin intermediate filaments (VIFs) reorganization affected CSs spreading only. The analysis of all three cytoskeleton systems contribution to spheroids intrinsic properties was performed for the first time.
Sigma-Aldrich
Phosphate buffered saline, tablet
Corning® 96 Well TC-Treated Microplates, size 96 wells, polystyrene plate, flat bottom, case of 50 (individually wrapped), sterile, lid (low evaporation)
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder