MilliporeSigma
  • Context-Specific Striatal Astrocyte Molecular Responses Are Phenotypically Exploitable.
Context-Specific Striatal Astrocyte Molecular Responses Are Phenotypically Exploitable.
Neuron (2020-10-22)
Xinzhu Yu, Jun Nagai, Maria Marti-Solano, Joselyn S Soto, Giovanni Coppola, M Madan Babu, Baljit S Khakh, Xinzhu Yu, Jun Nagai, Maria Marti-Solano, Joselyn S Soto, Giovanni Coppola, M Madan Babu, Baljit S Khakh
ABSTRACT
Astrocytes tile the central nervous system and are widely implicated in brain diseases, but the molecular mechanisms by which astrocytes contribute to brain disorders remain incompletely explored. By performing astrocyte gene expression analyses following 14 experimental perturbations of relevance to the striatum, we discovered that striatal astrocytes mount context-specific molecular responses at the level of genes, pathways, and upstream regulators. Through data mining, we also identified astrocyte pathways in Huntington's disease (HD) that were reciprocally altered with respect to the activation of striatal astrocyte G protein-coupled receptor (GPCR) signaling. Furthermore, selective striatal astrocyte stimulation of the Gi-GPCR pathway in vivo corrected several HD-associated astrocytic, synaptic, and behavioral phenotypes, with accompanying improvement of HD-associated astrocyte signaling pathways, including those related to synaptogenesis and neuroimmune functions. Overall, our data show that astrocytes are malleable, using context-specific responses that can be dissected molecularly and used for phenotypic benefit in brain disorders.
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Protease from Streptomyces griseus, BioReagent, DNase, RNase, and nickase, none detected (No RNase.)
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Anti-c-Fos Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Clozapine N-oxide