MilliporeSigma
  • MicroRNAs link estrogen receptor alpha status and Dicer levels in breast cancer.
MicroRNAs link estrogen receptor alpha status and Dicer levels in breast cancer.
Hormones & cancer (2011-07-16)
Dawn R Cochrane, Diana M Cittelly, Erin N Howe, Nicole S Spoelstra, Erin L McKinsey, Kelly LaPara, Anthony Elias, Douglas Yee, Jennifer K Richer
ABSTRACT
To identify microRNAs (miRNAs) associated with estrogen receptor (ESR1) status, we profiled luminal A, ESR1+ breast cancer cell lines versus triple negative (TN), which lack ERα, progesterone receptor and Her2/neu. Although two thirds of the differentially expressed miRNAs are higher in ESR1+ breast cancer cells, some miRNAs, such as miR-222/221 and miR-29a, are dramatically higher in ESR1- cells (∼100- and 16-fold higher, respectively). MiR-222/221 (which target ESR1 itself) and miR-29a are predicted to target the 3' UTR of Dicer1. Addition of these miRNAs to ESR1+ cells reduces Dicer protein, whereas antagonizing miR-222 in ESR1- cells increases Dicer protein. We demonstrate via luciferase reporter assays that these miRNAs directly target the Dicer1 3' UTR. In contrast, miR-200c, which promotes an epithelial phenotype, is 58-fold higher in the more well-differentiated ERα+ cells, and restoration of miR-200c to ERα- cells causes increased Dicer protein, resulting in increased levels of other mature miRNAs typically low in ESR1- cells. Together, our findings explain why Dicer is low in ERα negative breast cancers, since such cells express high miR-221/222 and miR-29a levels (which repress Dicer) and low miR-200c (which positively affect Dicer levels). Furthermore, we find that miR-7, which is more abundant in ERα+ cells and is estrogen regulated, targets growth factor receptors and signaling intermediates such as EGFR, IGF1R, and IRS-2. In summary, miRNAs differentially expressed in ERα+ versus ERα- breast cancers actively control some of the most distinguishing characteristics of the luminal A and TN subtypes, such as ERα itself, Dicer, and growth factor receptor levels.
Sigma-Aldrich
Anti-N-Cadherin Antibody, clone 13A9, culture supernatant, clone 13A9, Upstate®
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone V9, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid