MilliporeSigma
  • Lamin A-mediated nuclear lamina integrity is required for proper ciliogenesis.
Lamin A-mediated nuclear lamina integrity is required for proper ciliogenesis.
EMBO reports (2020-08-21)
Jia-Rong Fan, Li-Ru You, Won-Jing Wang, Wei-Syun Huang, Ching-Tung Chu, Ya-Hui Chi, Hong-Chen Chen
ABSTRACT
The primary cilium is a sensory organelle that receives specific signals from the extracellular environment important for vertebrate development and tissue homeostasis. Lamins, the major components of the nuclear lamina, are required to maintain the nuclear structure and are involved in most nuclear activities. In this study, we show that deficiency in lamin A/C causes defective ciliogenesis, accompanied by increased cytoplasmic accumulation of actin monomers and increased formation of actin filaments. Disruption of actin filaments by cytochalasin D rescues the defective ciliogenesis in lamin A/C-depleted cells. Moreover, lamin A/C-deficient cells display lower levels of nesprin 2 and defects in recruiting Arp2, myosin Va, and tau tubulin kinase 2 to the basal body during ciliogenesis. Collectively, our results uncover a functional link between nuclear lamina integrity and ciliogenesis and implicate the malfunction of primary cilia in the pathogenesis of laminopathy.
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-TTBK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture