MilliporeSigma
  • Ternary Cu(II) Complex with GHK Peptide and Cis-Urocanic Acid as a Potential Physiologically Functional Copper Chelate.
Ternary Cu(II) Complex with GHK Peptide and Cis-Urocanic Acid as a Potential Physiologically Functional Copper Chelate.
International journal of molecular sciences (2020-09-02)
Karolina Bossak-Ahmad, Marta D Wiśniewska, Wojciech Bal, Simon C Drew, Tomasz Frączyk
ABSTRACT
The tripeptide NH2-Gly-His-Lys-COOH (GHK), cis-urocanic acid (cis-UCA) and Cu(II) ions are physiological constituents of the human body and they co-occur (e.g., in the skin and the plasma). While GHK is known as Cu(II)-binding molecule, we found that urocanic acid also coordinates Cu(II) ions. Furthermore, both ligands create ternary Cu(II) complex being probably physiologically functional species. Regarding the natural concentrations of the studied molecules in some human tissues, together with the affinities reported here, we conclude that the ternary complex [GHK][Cu(II)][cis-urocanic acid] may be partly responsible for biological effects of GHK and urocanic acid described in the literature.
Sigma-Aldrich
cis-Urocanic acid, ≥98% (HPLC), solid (fluffy)
Sigma-Aldrich
Gly-His-Lys acetate salt, ≥97% (TLC)
Sigma-Aldrich
Imidazole, ≥99% (titration)