MilliporeSigma
  • First elucidation of the life cycle in the family Brachycladiidae (Digenea), parasites of marine mammals.
First elucidation of the life cycle in the family Brachycladiidae (Digenea), parasites of marine mammals.
International journal for parasitology (2020-07-15)
Georgii Kremnev, Anna Gonchar, Vladimir Krapivin, Olga Knyazeva, Darya Krupenko
ABSTRACT
Digeneans of the family Brachycladiidae are cosmopolitan parasites restricted to marine mammals. Their life cycles are unknown. Phylogenetically, Brachycladiidae are closely related to Acanthocolpidae, parasites of marine teleost fishes. Acanthocolpida typically possess three-host life cycles with gastropods of the superfamily Buccinoidea acting as the first intermediate hosts for most species, and either fishes or bivalves acting as the second intermediate hosts. A few species previously identified as Neophasis differ from other Acanthocolpidae in having naticid gastropods as first intermediate hosts, and both fishes and bivalves as second ones. We assumed that this may indicate an incorrect life cycle description and revised previous data on rediae and cercariae of Neophasis spp. from Cryptonatica affinis (Naticidae) and metacercariae from cardiid bivalves at the White Sea using molecular and morphological approaches. Sequence comparison showed that rediae and cercariae from C. affinis resembling some representatives of the genus Neophasis and metacercariae from bivalves resembling Neophasis oculata belong to the brachycladiid species Orthosplanchnus arcticus. Thus, the life cycle of O. arcticus proceeds as follows: seals serve as the definitive host, C. affinis as the first intermediate host and cardiid bivalves as the second. We found one more type of redia and cercaria in C. affinis which, by molecular evidence, also belongs to Brachycladiidae and is closely related to O. arcticus. Here we refer to them as Brachycladiidae gen. sp. 1 WS. We suggest that Brachycladiidae gen. sp. 1 WS may belong to either Orthosplanchnus or Odhneriella, with beluga whales possibly being the definitive host. Morphological features of O. arcticus and Brachycladiidae gen. sp. 1 WS cercariae are summarised and matched with published data on putatively brachycladiid cercariae. We compare and discuss the diversity of life cycle patterns among Brachycladiidae and Acanthocolpidae, and show that they differ not only in the type of definitive host, but also in both intermediate hosts.
Sigma-Aldrich
Phalloidin–Tetramethylrhodamine B isothiocyanate, sequence from Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Fluoroshield with DAPI, liquid
Sigma-Aldrich
Triton X-100, laboratory grade