MilliporeSigma
  • Antibodies to the RNA binding protein heterogeneous nuclear ribonucleoprotein A1 contribute to neuronal cell loss in an animal model of multiple sclerosis.
Antibodies to the RNA binding protein heterogeneous nuclear ribonucleoprotein A1 contribute to neuronal cell loss in an animal model of multiple sclerosis.
The Journal of comparative neurology (2019-12-25)
Cole D Libner, Hannah E Salapa, Catherine Hutchinson, Sangmin Lee, Michael C Levin
ABSTRACT
Neurodegeneration, including loss of neurons and axons, is a feature of progressive forms of multiple sclerosis (MS). The mechanisms underlying neurodegeneration are mostly unknown. Research implicates autoimmunity to nonmyelin self-antigens as important contributors to disease pathogenesis. Data from our lab implicate autoimmunity to the RNA binding protein (RBP) heterogeneous nuclear ribonucleoprotein A1 (hnRNP A1) as a possible mechanism of neurodegeneration in MS. MS patients make antibodies to hnRNP A1, which have been shown to lead to neuronal dysfunction in vitro. Using an animal model of MS, experimental autoimmune encephalomyelitis (EAE), we show here that injection of anti-hnRNP A1 antibodies, in contrast to control antibodies, resulted in worsened disease and increased neurodegeneration. We found a reduction of NeuN+ neuronal cell bodies in areas of the ventral gray matter of the spinal cord where anti-hnRNP A1 antibodies localized. Neurons displayed increased levels of hnRNP A1 nucleocytoplasmic mislocalization and stress granule formation, both markers of neuronal injury. Anti-hnRNP A1 antibodies were found to surround neuronal cell bodies and interact with CD68+ immune cells via Fc receptors. Additionally, anti-hnRNP A1 antibodies were found within neuronal cell bodies including those of the ventral spinocerebellar tract (VSCT), a tract previously shown to undergo neurodegeneration in anti-hnRNP A1 antibody injected EAE mice. Finally, both immune cells and neurons showed increased levels of inducible nitric oxide synthase, another indicator of cell damage. These findings suggest that autoimmunity to RBPs, such as hnRNP A1, play a role in neurodegeneration in EAE with important implications for the pathogenesis of MS.
Sigma-Aldrich
Mouse IgG2b Negative Control, clone GC198, Mouse IgG2b Negative Control Monoclonal Antibody validated for use in Flow Cytometry & Immunofluorescence.
Sigma-Aldrich
Anti-hnRNP A1 Antibody, clone 9H10, clone 9H10, from mouse
Sigma-Aldrich
DPX Mountant for histology, slide mounting medium
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-hnRNP A1 Antibody, clone 4B10, clone 4B10, from mouse
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Anti-Nitric Oxide Synthase, Inducible antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
DL-Cysteine, technical grade