MilliporeSigma
  • Netrin-1 promotes naive pluripotency through Neo1 and Unc5b co-regulation of Wnt and MAPK signalling.
Netrin-1 promotes naive pluripotency through Neo1 and Unc5b co-regulation of Wnt and MAPK signalling.
Nature cell biology (2020-04-02)
Aurélia Huyghe, Giacomo Furlan, Duygu Ozmadenci, Christina Galonska, Jocelyn Charlton, Xavier Gaume, Noémie Combémorel, Christina Riemenschneider, Nicolas Allègre, Jenny Zhang, Pauline Wajda, Nicolas Rama, Pauline Vieugué, Isabelle Durand, Marie Brevet, Nicolas Gadot, Thomas Imhof, Bradley J Merrill, Manuel Koch, Patrick Mehlen, Claire Chazaud, Alexander Meissner, Fabrice Lavial
ABSTRACT
In mouse embryonic stem cells (mESCs), chemical blockade of Gsk3α/β and Mek1/2 (2i) instructs a self-renewing ground state whose endogenous inducers are unknown. Here we show that the axon guidance cue Netrin-1 promotes naive pluripotency by triggering profound signalling, transcriptomic and epigenetic changes in mESCs. Furthermore, we demonstrate that Netrin-1 can substitute for blockade of Gsk3α/β and Mek1/2 to sustain self-renewal of mESCs in combination with leukaemia inhibitory factor and regulates the formation of the mouse pluripotent blastocyst. Mechanistically, we reveal how Netrin-1 and the balance of its receptors Neo1 and Unc5B co-regulate Wnt and MAPK pathways in both mouse and human ESCs. Netrin-1 induces Fak kinase to inactivate Gsk3α/β and stabilize β-catenin while increasing the phosphatase activity of a Ppp2r2c-containing Pp2a complex to reduce Erk1/2 activity. Collectively, this work identifies Netrin-1 as a regulator of pluripotency and reveals that it mediates different effects in mESCs depending on its receptor dosage, opening perspectives for balancing self-renewal and lineage commitment.
Sigma-Aldrich
GSK-3 Inhibitor XVI, CHIR99021, InSolution, ≥95%, 25 mM
Sigma-Aldrich
Alkaline Phosphatase Diethanolamine Activity Kit, sufficient for 100 tests
Sigma-Aldrich
TAT-CRE Recombinase, TAT-CRE Recombinase is a recombinant cell-permeant fusion cre-recombinase protein consisting of TAT sequence, a nuclear localization sequence (NLS) and it is known to catalyze the site specific recombination event between two loxP DNA sites.
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
MEK1/2 Inhibitor III, PD0325901, InSolution, ≥95%
Sigma-Aldrich
Anti-PP2A Antibody, C subunit, clone 1D6, clone 1D6, Upstate®, from mouse
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, ERK-2) antibody produced in rabbit, whole antiserum