MilliporeSigma
  • Hfq CLASH uncovers sRNA-target interaction networks linked to nutrient availability adaptation.
Hfq CLASH uncovers sRNA-target interaction networks linked to nutrient availability adaptation.
eLife (2020-05-02)
Ira Alexandra Iosub, Robert Willem van Nues, Stuart William McKellar, Karen Jule Nieken, Marta Marchioretto, Brandon Sy, Jai Justin Tree, Gabriella Viero, Sander Granneman
ABSTRACT
By shaping gene expression profiles, small RNAs (sRNAs) enable bacteria to efficiently adapt to changes in their environment. To better understand how Escherichia coli acclimatizes to nutrient availability, we performed UV cross-linking, ligation and sequencing of hybrids (CLASH) to uncover Hfq-associated RNA-RNA interactions at specific growth stages. We demonstrate that Hfq CLASH robustly captures bona fide RNA-RNA interactions. We identified hundreds of novel sRNA base-pairing interactions, including many sRNA-sRNA interactions and involving 3'UTR-derived sRNAs. We rediscovered known and identified novel sRNA seed sequences. The sRNA-mRNA interactions identified by CLASH have strong base-pairing potential and are highly enriched for complementary sequence motifs, even those supported by only a few reads. Yet, steady state levels of most mRNA targets were not significantly affected upon over-expression of the sRNA regulator. Our results reinforce the idea that the reproducibility of the interaction, not base-pairing potential, is a stronger predictor for a regulatory outcome.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane