MilliporeSigma
  • Intraflagellar Transport Complex B Proteins Regulate the Hippo Effector Yap1 during Cardiogenesis.
Intraflagellar Transport Complex B Proteins Regulate the Hippo Effector Yap1 during Cardiogenesis.
Cell reports (2020-07-23)
Marina Peralta, Laia Ortiz Lopez, Katerina Jerabkova, Tommaso Lucchesi, Benjamin Vitre, Dong Han, Laurent Guillemot, Chaitanya Dingare, Izabela Sumara, Nadia Mercader, Virginie Lecaudey, Benedicte Delaval, Sigolène M Meilhac, Julien Vermot
ABSTRACT
Cilia and the intraflagellar transport (IFT) proteins involved in ciliogenesis are associated with congenital heart diseases (CHDs). However, the molecular links between cilia, IFT proteins, and cardiogenesis are yet to be established. Using a combination of biochemistry, genetics, and live-imaging methods, we show that IFT complex B proteins (Ift88, Ift54, and Ift20) modulate the Hippo pathway effector YAP1 in zebrafish and mouse. We demonstrate that this interaction is key to restrict the formation of the proepicardium and the myocardium. In cellulo experiments suggest that IFT88 and IFT20 interact with YAP1 in the cytoplasm and functionally modulate its activity, identifying a molecular link between cilia-related proteins and the Hippo pathway. Taken together, our results highlight a noncanonical role for IFT complex B proteins during cardiogenesis and shed light on a mechanism of action for ciliary proteins in YAP1 regulation.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-AMOTL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Phosphate buffered saline, tablet
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
5-Propyl-2-thiouracil, ≥98%
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
Sigma-Aldrich
N-Phenylthiourea, ≥98%