MilliporeSigma
  • TDP-43 prevents retrotransposon activation in the Drosophila motor system through regulation of Dicer-2 activity.
TDP-43 prevents retrotransposon activation in the Drosophila motor system through regulation of Dicer-2 activity.
BMC biology (2020-07-06)
Giulia Romano, Raffaella Klima, Fabian Feiguin
ABSTRACT
Mutations in the small RNA-binding protein TDP-43 lead to the formation of insoluble cytoplasmic aggregates that have been associated with the onset and progression of amyotrophic lateral sclerosis (ALS), a neurodegenerative disorder affecting homeostasis of the motor system which is also characterized by aberrant expression of retrotransposable elements (RTEs). Although the TDP-43 function was shown to be required in the neurons and glia to maintain the organization of neuromuscular synapses and prevent denervation of the skeletal muscles, the molecular mechanisms involved in physiological dysregulation remain elusive. Here, we address this issue using a null mutation of the TDP-43 Drosophila homolog, TBPH. Using genome-wide gene expression profiles, we detected a strong upregulation of RTE expression in TBPH-null Drosophila heads, while the genetic rescue of the TDP-43 function reverted these modifications. Furthermore, we found that TBPH modulates the small interfering RNA (siRNA) silencing machinery responsible for RTE repression. Molecularly, we observed that TBPH regulates the expression levels of Dicer-2 by direct protein-mRNA interactions in vivo. Accordingly, the genetic or pharmacological recovery of Dicer-2 activity was sufficient to repress retrotransposon activation and promote motoneuron axonal wrapping and synaptic growth in TBPH-null Drosophila. We identified an upregulation of RTE expression in TBPH-null Drosophila heads and demonstrate that defects in the siRNA pathway lead to RTE upregulation and motoneuron degeneration. Our results describe a novel physiological role of endogenous TDP-43 in the prevention of RTE-induced neurological alterations through the modulation of Dicer-2 activity and the siRNA pathway.
Sigma-Aldrich
Lamivudine, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Abacavir sulfate, ≥98% (HPLC)
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®