MilliporeSigma
  • Naproxen Exhibits Broad Anti-influenza Virus Activity in Mice by Impeding Viral Nucleoprotein Nuclear Export.
Naproxen Exhibits Broad Anti-influenza Virus Activity in Mice by Impeding Viral Nucleoprotein Nuclear Export.
Cell reports (2019-05-09)
Weinan Zheng, Wenhui Fan, Shuang Zhang, Pengtao Jiao, Yingli Shang, Liang Cui, Madina Mahesutihan, Jing Li, Dayan Wang, George Fu Gao, Lei Sun, Wenjun Liu
ABSTRACT
Naproxen is a non-steroidal anti-inflammatory drug that has previously been shown to exert antiviral activity against influenza A virus by inhibiting nucleoprotein (NP) binding to RNA. Here, we show that naproxen is a potential broad, multi-mechanistic anti-influenza virus therapeutic, as it inhibits influenza B virus replication both in vivo and in vitro. The anti-influenza B virus activity of naproxen is more efficient than that of the commonly used neuraminidase inhibitor oseltamivir in mice. Furthermore, the NP of influenza B virus (BNP) has a higher binding affinity to naproxen than influenza A virus NP (ANP). Specifically, naproxen targets the NP at residues F209 (BNP) and Y148 (ANP). This interaction antagonizes the nuclear export of NP normally mediated by the host export protein CRM1. This study reveals a crucial mechanism of broad-spectrum anti-influenza virus activity of naproxen, suggesting that the existing drug naproxen may be used as an anti-influenza drug.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Millipore
ANTI-FLAG® M1 Agarose Affinity Gel
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution