MilliporeSigma
  • The switch-like expression of heme-regulated kinase 1 mediates neuronal proteostasis following proteasome inhibition.
The switch-like expression of heme-regulated kinase 1 mediates neuronal proteostasis following proteasome inhibition.
eLife (2020-04-25)
Beatriz Alvarez-Castelao, Susanne Tom Dieck, Claudia M Fusco, Paul Donlin-Asp, Julio D Perez, Erin M Schuman
ABSTRACT
We examined the feedback between the major protein degradation pathway, the ubiquitin-proteasome system (UPS), and protein synthesis in rat and mouse neurons. When protein degradation was inhibited, we observed a coordinate dramatic reduction in nascent protein synthesis in neuronal cell bodies and dendrites. The mechanism for translation inhibition involved the phosphorylation of eIF2α, surprisingly mediated by eIF2α kinase 1, or heme-regulated kinase inhibitor (HRI). Under basal conditions, neuronal expression of HRI is barely detectable. Following proteasome inhibition, HRI protein levels increase owing to stabilization of HRI and enhanced translation, likely via the increased availability of tRNAs for its rare codons. Once expressed, HRI is constitutively active in neurons because endogenous heme levels are so low; HRI activity results in eIF2α phosphorylation and the resulting inhibition of translation. These data demonstrate a novel role for neuronal HRI that senses and responds to compromised function of the proteasome to restore proteostasis.
Sigma-Aldrich
Anti-HRI Antibody, serum, Upstate®
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
7-Nitroindazole, ≥98%
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
Sigma-Aldrich
Heme Assay Kit, sufficient for 250 colorimetric tests
Sigma-Aldrich
(R)-MG132
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent