MilliporeSigma
  • Lysine Acetylation Reshapes the Downstream Signaling Landscape of Vav1 in Lymphocytes.
Lysine Acetylation Reshapes the Downstream Signaling Landscape of Vav1 in Lymphocytes.
Cells (2020-03-08)
Sonia Rodríguez-Fdez, Lucía Fernández-Nevado, L Francisco Lorenzo-Martín, Xosé R Bustelo
ABSTRACT
Vav1 works both as a catalytic Rho GTPase activator and an adaptor molecule. These functions, which are critical for T cell development and antigenic responses, are tyrosine phosphorylation-dependent. However, it is not known whether other posttranslational modifications can contribute to the regulation of the biological activity of this protein. Here, we show that Vav1 becomes acetylated on lysine residues in a stimulation- and SH2 domain-dependent manner. Using a collection of both acetylation- and deacetylation-mimicking mutants, we show that the acetylation of four lysine residues (Lys222, Lys252, Lys587, and Lys716) leads to the downmodulation of the adaptor function of Vav1 that triggers the stimulation of the nuclear factor of activated T cells (NFAT). These sites belong to two functional subclasses according to mechanistic criteria. We have also unveiled additional acetylation sites potentially involved in either the stimulation (Lys782) or the downmodulation (Lys335, Lys374) of specific Vav1-dependent downstream responses. Collectively, these results indicate that Nε-lysine acetylation can play variegated roles in the regulation of Vav1 signaling. Unlike the case of the tyrosine phosphorylation step, this new regulatory layer is not conserved in other Vav family paralogs.
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Nα-Acetyl-L-lysine
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
DEAE-Dextran hydrochloride, powder
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®
Sigma-Aldrich
Anti-Acetyl Lysine Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-CD3 Mouse mAb (UCHT1), liquid, >95% (immunoglobulin, SDS-PAGE), clone UCHT1
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Triton X-100, laboratory grade