MilliporeSigma
  • Critical Roles of Translation Initiation and RNA Uridylation in Endogenous Retroviral Expression and Neural Differentiation in Pluripotent Stem Cells.
Critical Roles of Translation Initiation and RNA Uridylation in Endogenous Retroviral Expression and Neural Differentiation in Pluripotent Stem Cells.
Cell reports (2020-06-04)
Kazutoshi Takahashi, Daeun Jeong, Songnan Wang, Megumi Narita, Xuemei Jin, Mio Iwasaki, Samuel D Perli, Bruce R Conklin, Shinya Yamanaka
ABSTRACT
Previous studies have suggested that the loss of the translation initiation factor eIF4G1 homolog NAT1 induces excessive self-renewability of naive pluripotent stem cells (PSCs); yet the role of NAT1 in the self-renewal and differentiation of primed PSCs is still unclear. Here, we generate a conditional knockout of NAT1 in primed PSCs and use the cells for the functional analyses of NAT1. Our results show that NAT1 is required for the self-renewal and neural differentiation of primed PSCs. In contrast, NAT1 deficiency in naive pluripotency attenuates the differentiation to all cell types. We also find that NAT1 is involved in efficient protein expression of an RNA uridyltransferase, TUT7. TUT7 is involved in the neural differentiation of primed PSCs via the regulation of human endogenous retrovirus accumulation. These data demonstrate the essential roles of NAT1 and TUT7 in the precise transition of stem cell fate.
Sigma-Aldrich
Magnesium methyl carbonate solution, 2.0 M in DMF
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Leukemia Inhibitory Factor human, 10 µg, human recombinant LIF protein, expressed in E. coli, suitable for stem cell culture
Sigma-Aldrich
Alkaline Phosphatase Detection Kit, This Alkaline Phosphatase Detection Kit is a specific & sensitive tool for the phenotypic assessment of Embryonic Stem (ES) cell differentiation by the determination of AP activity.
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Avanti
(2S)-OMPT, Avanti Research - A Croda Brand
Sigma-Aldrich
Anti-Nuclei Antibody, clone 3E1.3, clone 3E1.3, Chemicon®, from mouse
Millipore
Accutase cell detachment solution, A cell detachment solution of proteolytic & collagenolytic enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.
Sigma-Aldrich
Anti-ZCCHC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
MG-132, InSolution, ≥98%, 10 mM, reversible proteasome inhibitor
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.