MilliporeSigma
A salvage pathway maintains highly functional respiratory complex I.
Nature communications (2020-04-04)
Karolina Szczepanowska, Katharina Senft, Juliana Heidler, Marija Herholz, Alexandra Kukat, Michaela Nicole Höhne, Eduard Hofsetz, Christina Becker, Sophie Kaspar, Heiko Giese, Klaus Zwicker, Sergio Guerrero-Castillo, Linda Baumann, Johanna Kauppila, Anastasia Rumyantseva, Stefan Müller, Christian K Frese, Ulrich Brandt, Jan Riemer, Ilka Wittig, Aleksandra Trifunovic
ABSTRACT
Regulation of the turnover of complex I (CI), the largest mitochondrial respiratory chain complex, remains enigmatic despite huge advancement in understanding its structure and the assembly. Here, we report that the NADH-oxidizing N-module of CI is turned over at a higher rate and largely independently of the rest of the complex by mitochondrial matrix protease ClpXP, which selectively removes and degrades damaged subunits. The observed mechanism seems to be a safeguard against the accumulation of dysfunctional CI arising from the inactivation of the N-module subunits due to attrition caused by its constant activity under physiological conditions. This CI salvage pathway maintains highly functional CI through a favorable mechanism that demands much lower energetic cost than de novo synthesis and reassembly of the entire CI. Our results also identify ClpXP activity as an unforeseen target for therapeutic interventions in the large group of mitochondrial diseases characterized by the CI instability.
Sigma-Aldrich
Auranofin, ≥98% (HPLC)
Sigma-Aldrich
Myxothiazol, from Myxococcus fulvus Mx f85, ≥98% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Anti-CLPX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Calnexin, C-Terminal (575-593) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)