MilliporeSigma
  • Phosphorylation of TFCP2L1 by CDK1 is required for stem cell pluripotency and bladder carcinogenesis.
Phosphorylation of TFCP2L1 by CDK1 is required for stem cell pluripotency and bladder carcinogenesis.
EMBO molecular medicine (2019-11-12)
Jinbeom Heo, Byeong-Joo Noh, Seungun Lee, Hye-Yeon Lee, YongHwan Kim, Jisun Lim, Hyein Ju, Hwan Yeul Yu, Chae-Min Ryu, Peter Cw Lee, Hwangkyo Jeong, Yumi Oh, Kyunggon Kim, Sang-Yeob Kim, Jaekyoung Son, Bumsik Hong, Jong Soo Kim, Yong Mee Cho, Dong-Myung Shin
ABSTRACT
Molecular programs involved in embryogenesis are frequently upregulated in oncogenic dedifferentiation and metastasis. However, their precise roles and regulatory mechanisms remain elusive. Here, we showed that CDK1 phosphorylation of TFCP2L1, a pluripotency-associated transcription factor, orchestrated pluripotency and cell-cycling in embryonic stem cells (ESCs) and was aberrantly activated in aggressive bladder cancers (BCs). In murine ESCs, the protein interactome and transcription targets of Tfcp2l1 indicated its involvement in cell cycle regulation. Tfcp2l1 was phosphorylated at Thr177 by Cdk1, which affected ESC cell cycle progression, pluripotency, and differentiation. The CDK1-TFCP2L1 pathway was activated in human BC cells, stimulating their proliferation, self-renewal, and invasion. Lack of TFCP2L1 phosphorylation impaired the tumorigenic potency of BC cells in a xenograft model. In patients with BC, high co-expression of TFCP2L1 and CDK1 was associated with unfavorable clinical characteristics including tumor grade, lymphovascular and muscularis propria invasion, and distant metastasis and was an independent prognostic factor for cancer-specific survival. These findings demonstrate the molecular and clinical significance of CDK1-mediated TFCP2L1 phosphorylation in stem cell pluripotency and in the tumorigenic stemness features associated with BC progression.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Klf2 Antibody, serum, from rabbit