MilliporeSigma
Loss of Phd2 cooperates with BRAFV600E to drive melanomagenesis.
Nature communications (2018-12-24)
Shujing Liu, Gao Zhang, Jianping Guo, Xiang Chen, Jingce Lei, Kan Ze, Liyun Dong, Xiangpeng Dai, Yang Gao, Daisheng Song, Brett L Ecker, Ruifeng Yang, Caitlin Feltcher, Kai Peng, Cheng Feng, Hui Chen, Rebecca X Lee, Heddy Kerestes, Jingwen Niu, Suresh Kumar, Weiting Xu, Jie Zhang, Zhi Wei, James S Martin, Xiaoming Liu, Gordon Mills, Yiling Lu, Wei Guo, Lunquan Sun, Lin Zhang, Ashani Weeraratna, Meenhard Herlyn, Wenyi Wei, Frank S Lee, Xiaowei Xu
ABSTRACT
Prolyl hydroxylase domain protein 2 (PHD2) is a well-known master oxygen sensor. However, the role of PHD2 in tumor initiation remains controversial. We find that during the transition of human nevi to melanoma, the expression of PHD2 protein is significantly decreased and lower expression PHD2 in melanoma is associated with worse clinical outcome. Knockdown of PHD2 leads to elevated Akt phosphorylation in human melanocytes. Mice with conditional melanocyte-specific expression of Phd2lox/lox (Tyr::CreER;Phd2lox/lox) fail to develop pigmented lesions. However, deletion of Phd2 in combination with expression of BRafV600E in melanocytes (Tyr::CreER;Phd2lox/lox;BRafCA) leads to the development of melanoma with 100% penetrance and frequent lymph node metastasis. Analysis of tumor tissues using reverse phase protein arrays demonstrates that Phd2 deletion activates the AKT-mTOR-S6 signaling axis in the recovered tumors. These data indicate that PHD2 is capable of suppressing tumor initiation largely mediated through inhibiting of the Akt-mTOR signaling pathway in the melanocyte lineage.
Sigma-Aldrich
Anti-VEGF-A Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Roche
Cell Proliferation Kit I (MTT)
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid