MilliporeSigma
  • Quantitative analysis of protein-RNA interactions by gel mobility shift.
Quantitative analysis of protein-RNA interactions by gel mobility shift.
Methods in molecular biology (Clifton, N.J.) (2008-11-05)
Sean P Ryder, Michael I Recht, James R Williamson
ABSTRACT
The gel mobility shift assay is routinely used to visualize protein-RNA interactions. Its power resides in the ability to resolve free from bound RNA with high resolution in a gel matrix. We review the quantitative application of this approach to elucidate thermodynamic properties of protein-RNA complexes. Assay designs for titration, competition, and stoichiometry experiments are presented for two unrelated model complexes.
Sigma-Aldrich
IGEPAL® CA-630, Molecular Biology