MilliporeSigma
p53 Represses the Mevalonate Pathway to Mediate Tumor Suppression.
Cell (2018-12-26)
Sung-Hwan Moon, Chun-Hao Huang, Shauna L Houlihan, Kausik Regunath, William A Freed-Pastor, John P Morris, Darjus F Tschaharganeh, Edward R Kastenhuber, Anthony M Barsotti, Rachel Culp-Hill, Wen Xue, Yu-Jui Ho, Timour Baslan, Xiang Li, Allison Mayle, Elisa de Stanchina, Lars Zender, David R Tong, Angelo D'Alessandro, Scott W Lowe, Carol Prives
ABSTRACT
There are still gaps in our understanding of the complex processes by which p53 suppresses tumorigenesis. Here we describe a novel role for p53 in suppressing the mevalonate pathway, which is responsible for biosynthesis of cholesterol and nonsterol isoprenoids. p53 blocks activation of SREBP-2, the master transcriptional regulator of this pathway, by transcriptionally inducing the ABCA1 cholesterol transporter gene. A mouse model of liver cancer reveals that downregulation of mevalonate pathway gene expression by p53 occurs in premalignant hepatocytes, when p53 is needed to actively suppress tumorigenesis. Furthermore, pharmacological or RNAi inhibition of the mevalonate pathway restricts the development of murine hepatocellular carcinomas driven by p53 loss. Like p53 loss, ablation of ABCA1 promotes murine liver tumorigenesis and is associated with increased SREBP-2 maturation. Our findings demonstrate that repression of the mevalonate pathway is a crucial component of p53-mediated liver tumor suppression and outline the mechanism by which this occurs.
Sigma-Aldrich
Nutlin-3, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-ABCA1 Antibody, clone AB.H10, clone AB.H10, Chemicon®, from mouse
Sigma-Aldrich
Calpain Inhibitor I, ≥97% (TLC), powder
Roche
Dispase® II (neutral protease, grade II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
(±)-Mevalonic acid 5-phosphate lithium salt hydrate, 95% (TLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
IgG from mouse serum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder