MilliporeSigma
ERG Controls B Cell Development by Promoting Igh V-to-DJ Recombination.
Cell reports (2019-11-28)
Elisabeth Søndergaard, Alexander Rauch, Magali Michaut, Nicolas Rapin, Matilda Rehn, Anna S Wilhelmson, Alessandro Camponeschi, Marie S Hasemann, Frederik O Bagger, Johan Jendholm, Kasper J Knudsen, Susanne Mandrup, Inga-Lill Mårtensson, Bo T Porse
ABSTRACT
B cell development depends on the coordinated expression and cooperation of several transcription factors. Here we show that the transcription factor ETS-related gene (ERG) is crucial for normal B cell development and that its deletion results in a substantial loss of bone marrow B cell progenitors and peripheral B cells, as well as a skewing of splenic B cell populations. We find that ERG-deficient B lineage cells exhibit an early developmental block at the pre-B cell stage and proliferate less. The cells fail to express the immunoglobulin heavy chain due to inefficient V-to-DJ recombination, and cells that undergo recombination display a strong bias against incorporation of distal V gene segments. Furthermore, antisense transcription at PAX5-activated intergenic repeat (PAIR) elements, located in the distal region of the Igh locus, depends on ERG. These findings show that ERG serves as a critical regulator of B cell development by ensuring efficient and balanced V-to-DJ recombination.
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Dipeptidyl Peptidase III human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Saponin, used as non-ionic surfactant
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein